Assay Details
| Gene Symbol: Trim24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272064.1 | NP_001258993.1 | 17 - 18 | 2989 | 135 | |
| NM_001272076.1 | NP_001259005.1 | 17 - 18 | 2671 | 135 | ||
| NM_145076.4 | NP_659542.3 | 17 - 18 | 3091 | 135 | ||
| GenBank mRNA | AB221637.1 | - | 17 - 18 | 2791 | 135 | |
| AK133144.1 | - | 17 - 18 | 2671 | 135 | ||
| AK166538.1 | - | 13 - 14 | 2048 | 135 | ||
| BC025482.1 | - | 8 - 9 | 1186 | 135 | ||
| BC056959.1 | - | 17 - 18 | 3069 | 5698079 | 135 | |
| S78219.1 | - | 17 - 18 | 2989 | 135 | ||
| S78221.1 | - | 17 - 18 | 3091 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim24
Genomic Map