| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700014299.mm.1 |
TC0700001710.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700014299.mm.1 |
NM_001177977.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014299.mm.1 |
NM_001177977.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014299.mm.1 |
NM_001177978.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014299.mm.1 |
NM_001177978.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014299.mm.1 |
NM_146197.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014299.mm.1 |
NM_146197.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014302.mm.1 |
TC0700001710.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700014302.mm.1 |
NM_001177977.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014302.mm.1 |
NM_001177977.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014302.mm.1 |
NM_001177978.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014302.mm.1 |
NM_001177978.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014302.mm.1 |
NM_146197.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014302.mm.1 |
NM_146197.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014301.mm.1 |
TC0700001710.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700014301.mm.1 |
NM_001177977.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014301.mm.1 |
NM_001177977.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014301.mm.1 |
NM_001177978.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014301.mm.1 |
NM_001177978.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014301.mm.1 |
NM_146197.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014301.mm.1 |
NM_146197.4 |
Refseq |