Assay Details
| Gene Symbol: Tubgcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286007.1 | NP_001272936.1 | 11 - 12 | 2084 | 60 | |
| NM_001286009.1 | NP_001272938.1 | 11 - 12 | 2084 | 60 | ||
| NM_001286011.1 | NP_001272940.1 | 10 - 11 | 1851 | 60 | ||
| NM_133755.2 | NP_598516.2 | 11 - 12 | 2011 | 60 | ||
| GenBank mRNA | AK006233.1 | - | 1 - 2 | 115 | 60 | |
| AK033157.1 | - | 11 - 12 | 1801 | 60 | ||
| AK045084.1 | - | 11 - 12 | 1800 | 60 | ||
| AK146771.1 | - | 11 - 12 | 2006 | 60 | ||
| AK153511.1 | - | 11 - 12 | 1932 | 60 | ||
| AK158060.1 | - | 11 - 12 | 1800 | 60 | ||
| AK167924.1 | - | 11 - 12 | 1906 | 60 | ||
| AK194815.1 | - | 2 - 3 | 277 | 60 | ||
| BC012519.1 | - | 11 - 12 | 1939 | 3601589 | 60 | |
| BC025564.1 | - | 10 - 11 | 1773 | 60 | ||
| BC025582.1 | - | 11 - 12 | 1900 | 5324981 | 60 | |
| BC027034.1 | - | 10 - 11 | 2285 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tubgcp2
Genomic Map