Assay Details
| Gene Symbol: Kat8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026370.1 | NP_080646.1 | 5 - 6 | 697 | 126 | |
| GenBank mRNA | AK003264.1 | - | 5 - 6 | 697 | 126 | |
| AK017161.1 | - | 4 - 5 | 443 | 126 | ||
| AK018002.1 | - | 5 - 6 | 694 | 125 | ||
| AK132000.1 | - | 5 - 6 | 698 | 126 | ||
| AK146710.1 | - | 5 - 6 | 699 | 126 | ||
| BC036284.1 | - | 5 - 6 | 705 | 5136304 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kat8
Genomic Map