Assay Details
| Gene Symbol: Kat8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026370.1 | NP_080646.1 | 7 - 8 | 930 | 62 | |
| GenBank mRNA | AK003264.1 | - | 7 - 8 | 930 | 62 | |
| AK008418.1 | - | 7 - 8 | 917 | 62 | ||
| AK017161.1 | - | 6 - 7 | 676 | 62 | ||
| AK018002.1 | - | 7 - 8 | 926 | 62 | ||
| AK132000.1 | - | 7 - 8 | 931 | 62 | ||
| AK146710.1 | - | 7 - 8 | 932 | 62 | ||
| BC036284.1 | - | 7 - 8 | 938 | 5136304 | 62 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kat8
Genomic Map