Assay Details
| Gene Symbol: Kat8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026370.1 | NP_080646.1 | 9 - 10 | 1172 | 84 | |
| GenBank mRNA | AK003264.1 | - | 9 - 10 | 1172 | 84 | |
| AK008418.1 | - | 9 - 10 | 1159 | 84 | ||
| AK017161.1 | - | 8 - 9 | 918 | 84 | ||
| AK018002.1 | - | 9 - 10 | 1168 | 84 | ||
| AK132000.1 | - | 9 - 10 | 1173 | 84 | ||
| AK146710.1 | - | 9 - 10 | 1174 | 84 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kat8
Genomic Map