Assay Details
| Gene Symbol: Tpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013861.3 | NP_038889.1 | 2 - 3 | 92 | 72 | |
| GenBank mRNA | AB027568.1 | - | 2 - 3 | 93 | 72 | |
| AK034521.1 | - | 102 | 72 | |||
| AK035044.1 | - | 2 - 3 | 93 | 72 | ||
| AK039131.1 | - | 2 - 3 | 137 | 72 | ||
| AK052225.1 | - | 2 - 3 | 83 | 72 | ||
| AK087844.1 | - | 2 - 3 | 196 | 72 | ||
| AK136486.1 | - | 2 - 3 | 93 | 72 | ||
| BC015246.1 | - | 2 - 3 | 109 | 4221866 | 72 | |
| BC023354.1 | - | 2 - 3 | 68 | 5253545 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tpk1
Genomic Map