Assay Details
| Gene Symbol: Tom1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039092.3 | NP_001034181.1 | 5 - 6 | 595 | 60 | |
| NM_001039093.1 | NP_001034182.1 | 5 - 6 | 595 | 60 | ||
| NM_153080.2 | NP_694720.2 | 5 - 6 | 595 | 60 | ||
| GenBank mRNA | AF467887.1 | - | 5 - 6 | 575 | 60 | |
| AK043095.1 | - | 5 - 6 | 575 | 60 | ||
| AK054531.1 | - | 5 - 6 | 563 | 60 | ||
| AK166040.1 | - | 5 - 6 | 595 | 60 | ||
| BC062947.1 | - | 5 - 6 | 559 | 30536491 | 60 | |
| BC127266.1 | - | 5 - 6 | 561 | 30357295 | 60 | |
More Information
Set Membership: |
|

Tom1l2
Genomic Map