Assay Details
Gene Symbol: Prl8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028477.2 | NP_082753.1 | 2 - 3 | 272 | 89 | |
GenBank mRNA | AF525158.1 | - | 2 - 3 | 229 | 89 | |
AK005485.1 | - | 2 - 3 | 273 | 89 | ||
AK014414.1 | - | 2 - 3 | 273 | 89 | ||
BC109185.1 | - | 2 - 3 | 271 | 40050251 | 89 | |
BC109186.1 | - | 2 - 3 | 271 | 40050253 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
