Assay Details
Gene Symbol: Tspan17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028841.3 | NP_083117.2 | 3 - 4 | 473 | 56 | |
GenBank mRNA | AK008761.1 | - | 3 - 4 | 418 | 56 | |
AK166272.1 | - | 3 - 4 | 476 | 56 | ||
AK171691.1 | - | 3 - 4 | 422 | 56 | ||
BC010346.1 | - | 3 - 4 | 418 | 2811935 | 56 | |
BY734818.1 | - | 3 - 4 | 473 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
