Assay Details
| Gene Symbol: Mvp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080638.3 | NP_542369.2 | 7 - 8 | 1009 | 82 | |
| GenBank mRNA | AF210456.1 | - | 7 - 8 | 1011 | 82 | |
| AK088046.1 | - | 7 - 8 | 994 | 82 | ||
| AK152534.1 | - | 7 - 8 | 984 | 82 | ||
| AK167899.1 | - | 7 - 8 | 985 | 82 | ||
| AK168100.1 | - | 7 - 8 | 985 | 82 | ||
| BC006709.1 | - | 7 - 8 | 974 | 3584439 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mvp
Genomic Map