Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000011014.mm.1 |
NM_008080.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011014.mm.1 |
NM_008080.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011014.mm.1 |
NM_027739.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011014.mm.1 |
NM_027739.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011014.mm.1 |
NR_045166.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011014.mm.1 |
NR_045166.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011014.mm.1 |
TC1000001551.mm.1 |
MTA TC |