Assay Details
| Gene Symbol: Cdkal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004490.1 | - | 2 - 3 | 253 | 149 | |
| AK082629.1 | - | 13 - 14 | 1426 | 149 | ||
| BC016073.1 | - | 13 - 14 | 1367 | 4482130 | 149 | |
More Information
Set Membership: |
|

Cdkal1
Genomic Map