Assay Details
| Gene Symbol: Cdkal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004490.1 | - | 3 - 4 | 330 | 109 | |
| AK082629.1 | - | 14 - 15 | 1503 | 109 | ||
| BC016073.1 | - | 14 - 15 | 1444 | 4482130 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdkal1
Genomic Map