Assay Details
| Gene Symbol: Yap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171147.1 | NP_001164618.1 | 4 - 5 | 960 | 132 | |
| NM_009534.3 | NP_033560.1 | 4 - 5 | 960 | 132 | ||
| GenBank mRNA | AK182200.1 | - | 2 - 3 | 144 | 132 | |
| BC014733.1 | - | 4 - 5 | 957 | 4239820 | 132 | |
| BC039125.1 | - | 4 - 5 | 815 | 2649070 | 132 | |
| BC094313.1 | - | 4 - 5 | 948 | 5697882 | 132 | |
| X80508.1 | - | 4 - 5 | 952 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Yap1
Genomic Map