Assay Details
| Gene Symbol: Dcun1d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029775.2 | NP_084051.1 | 2 - 3 | 410 | 61 | |
| GenBank mRNA | AK012764.1 | - | 2 - 3 | 423 | 61 | |
| AK013933.1 | - | 2 - 3 | 428 | 61 | ||
| AK014737.1 | - | 2 - 3 | 452 | 61 | ||
| AK134862.1 | - | 457 | 61 | |||
| BC020048.1 | - | 2 - 3 | 446 | 61 | ||
| BC020089.1 | - | 2 - 3 | 410 | 4013755 | 61 | |
| BC049641.1 | - | 599 | 61 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dcun1d5
Genomic Map