Assay Details
| Gene Symbol: Ascc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199187.2 | NP_001186116.1 | 4 - 5 | 424 | 59 | |
| NM_026937.3 | NP_081213.1 | 4 - 5 | 437 | 59 | ||
| GenBank mRNA | AK007519.1 | - | 4 - 5 | 421 | 59 | |
| AK053811.1 | - | 4 - 5 | 438 | 59 | ||
| AK139944.1 | - | 4 - 5 | 424 | 59 | ||
| AK144614.1 | - | 4 - 5 | 398 | 59 | ||
| AK171958.1 | - | 4 - 5 | 398 | 59 | ||
| AK201964.1 | - | 3 - 4 | 281 | 59 | ||
| BC030905.1 | - | 4 - 5 | 403 | 4503445 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ascc1
Genomic Map