Assay Details
| Gene Symbol: Ascc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199187.2 | NP_001186116.1 | 7 - 8 | 854 | 86 | |
| NM_026937.3 | NP_081213.1 | 7 - 8 | 867 | 86 | ||
| GenBank mRNA | AK007519.1 | - | 7 - 8 | 851 | 86 | |
| AK139944.1 | - | 7 - 8 | 854 | 86 | ||
| AK171958.1 | - | 7 - 8 | 828 | 86 | ||
| BC030905.1 | - | 7 - 8 | 833 | 4503445 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ascc1
Genomic Map