Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000003561.mm.1 |
NM_023530.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003561.mm.1 |
NM_023530.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003561.mm.1 |
TC1000000534.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003562.mm.1 |
NM_023530.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003562.mm.1 |
NM_023530.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003562.mm.1 |
TC1000000534.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003563.mm.1 |
NM_023530.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003563.mm.1 |
NM_023530.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003563.mm.1 |
TC1000000534.mm.1 |
MTA TC |