Assay Details
| Gene Symbol: Cfd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291915.1 | NP_001278844.1 | 4 - 5 | 688 | 76 | |
| NM_013459.3 | NP_038487.1 | 4 - 5 | 691 | 76 | ||
| GenBank mRNA | AK143817.1 | - | 4 - 5 | 635 | 76 | |
| BC138778.1 | - | 4 - 5 | 691 | 8861800 | 76 | |
| BC138779.1 | - | 4 - 5 | 691 | 8861801 | 76 | |
| BC145412.1 | - | 4 - 5 | 688 | 9053970 | 76 | |
| BE136302.1 | - | 1 - 2 | 76 | |||
| M11768.1 | - | 4 - 5 | 632 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P156955 | M11768.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P156955 | BE136302.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P156955 | NM_013459.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P156955 | AK143817.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P156955 | M11768.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P156955 | BE136302.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P156955 | NM_013459.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P156955 | AK143817.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 762696 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175484 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P156955 | M11768.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P156955 | BE136302.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P156955 | NM_013459.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P156955 | AK143817.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P156955 | M11768.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P156955 | BE136302.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P156955 | NM_013459.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P156955 | AK143817.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P156955 | M11768.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P156955 | BE136302.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P156955 | NM_013459.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P156955 | AK143817.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P156955 | M11768.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P156955 | BE136302.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P156955 | NM_013459.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P156955 | AK143817.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27279536 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175484 | - | - |
| Whole Mouse Genome G4122A | A_51_P156955 | M11768.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P156955 | BE136302.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P156955 | NM_013459.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P156955 | AK143817.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P156955 | M11768.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P156955 | BE136302.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P156955 | NM_013459.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P156955 | AK143817.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105658 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102086 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175484 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08504565 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08504584 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08504563 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08504562 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08504556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27279495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08504565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08793098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08504584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08504559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27279521 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10364546 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364547 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364545 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364544 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364543 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417867_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10364542 | - | - |
| Mouse Genome 430 2.0 Array | 1417867_at | - | - |
| Mouse Genome 430 2.0 Array | 1417867_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11537_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11537_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000006035.mm.1 | NM_013459.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000006035.mm.1 | TC1000000824.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000006036.mm.1 | NM_013459.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000006036.mm.1 | TC1000000824.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417867_PM_at | - | - |
| Mouse Expression Set 430 | 1417867_at | - | - |
| Murine Genome U74v2 Set | 99671_at | - | - |
| Murine Genome U74v2 Set | 99671_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417867_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417867_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417867_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417867_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 916908 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2320736 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2320736 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002320736 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230957 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230957 | - | - |
| mouse-6 v1.1 (Illumina) | scl38703.5.1_6-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2320736 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38703.5.1_6-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2320736 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2835423 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2835423 | - | - |

Cfd
Genomic Map