Assay Details
| Gene Symbol: Agpat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053014.3 | NP_443747.2 | 7 - 8 | 946 | 112 | |
| GenBank mRNA | AB377215.1 | - | 5 - 6 | 762 | 112 | |
| AK008965.1 | - | 7 - 8 | 1102 | 112 | ||
| AK015906.1 | - | 6 - 7 | 874 | 112 | ||
| AK030607.1 | - | 5 - 6 | 738 | 112 | ||
| AK075715.1 | - | 7 - 8 | 946 | 112 | ||
| AK076414.1 | - | 6 - 7 | 915 | 112 | ||
| AY167588.1 | - | 5 - 6 | 784 | 112 | ||
| BC033444.1 | - | 5 - 6 | 624 | 112 | ||
| BC052382.1 | - | 6 - 7 | 837 | 5703571 | 112 | |
| BC058519.1 | - | 6 - 7 | 836 | 5703571 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agpat3
Genomic Map