Assay Details
Gene Symbol: Ddx21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019553.2 | NP_062426.2 | 7 - 8 | 1536 | 145 | |
GenBank mRNA | AF159131.3 | - | 7 - 8 | 1497 | 145 | |
AK010281.1 | - | 7 - 8 | 1416 | 145 | ||
AK145578.1 | - | 7 - 8 | 1520 | 145 | ||
AK160095.1 | - | 7 - 8 | 1510 | 145 | ||
BC030895.1 | - | 6 - 7 | 968 | 145 | ||
BC043655.1 | - | 7 - 8 | 1508 | 5363948 | 145 | |
BC059237.1 | - | 7 - 8 | 1496 | 6412782 | 145 | |
BC060220.1 | - | 7 - 8 | 1488 | 145 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
