Assay Details
| Gene Symbol: Slc25a16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175194.2 | NP_780403.1 | 7 - 8 | 891 | 73 | |
| GenBank mRNA | AK030865.1 | - | 7 - 8 | 891 | 73 | |
| BC025015.1 | - | 7 - 8 | 802 | 73 | ||
| BC138983.1 | - | 7 - 8 | 813 | 8862005 | 73 | |
| BC138984.1 | - | 7 - 8 | 813 | 8862006 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a16
Genomic Map