Assay Details
Gene Symbol: Med16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163276.1 | NP_001156748.1 | 9 - 10 | 1669 | 71 | |
NM_198107.2 | NP_932775.2 | 9 - 10 | 1672 | 71 | ||
GenBank mRNA | AK042340.1 | - | 9 - 10 | 1634 | 71 | |
AK149345.1 | - | 9 - 10 | 1653 | 71 | ||
BC057056.1 | - | 9 - 10 | 1626 | 6813010 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
