Assay Details
| Gene Symbol: Pald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013753.2 | NP_038781.2 | 6 - 7 | 1061 | 88 | |
| GenBank mRNA | AK088577.1 | - | 6 - 7 | 897 | 88 | |
| AK146489.1 | - | 6 - 7 | 1006 | 88 | ||
| BC137695.1 | - | 5 - 6 | 825 | 8860714 | 88 | |
| BC144847.1 | - | 5 - 6 | 831 | 9053395 | 88 | |
More Information
Set Membership: |
|

Pald1
Genomic Map