Assay Details
Gene Symbol: Nt5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011851.4 | NP_035981.1 | 4 - 5 | 1036 | 159 | |
GenBank mRNA | AK028723.1 | - | 4 - 5 | 1006 | 159 | |
AK029979.1 | - | 4 - 5 | 1006 | 159 | ||
AK128997.1 | - | 4 - 5 | 1005 | 159 | ||
AK154614.1 | - | 4 - 5 | 1013 | 159 | ||
BC119268.1 | - | 4 - 5 | 980 | 8734017 | 159 | |
BC119270.1 | - | 4 - 5 | 980 | 8734019 | 159 | |
L12059.1 | - | 4 - 5 | 959 | 159 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
