Assay Details
Gene Symbol: Mbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF072248.1 | - | 2 - 3 | 522 | 57 | |
BC038264.1 | - | 2 - 3 | 323 | 5250237 | 57 | |
BQ042794.1 | - | 1 - 2 | 90 | 57 | ||
CK625083.1 | - | 2 - 3 | 342 | 57 | ||
CX208033.1 | - | 2 - 3 | 414 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
