Assay Details
| Gene Symbol: Prss57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042710.1 | NP_001036175.1 | 1 - 2 | 140 | 88 | |
| GenBank mRNA | AK153740.1 | - | 1 - 2 | 107 | 88 | |
| AK158376.1 | - | 1 - 2 | 140 | 88 | ||
| BC116380.2 | - | 2 - 3 | 290 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prss57
Genomic Map