Assay Details
Gene Symbol: Gap43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008083.2 | NP_032109.1 | 1 - 2 | 185 | 82 | |
GenBank mRNA | AK031908.1 | - | 1 - 2 | 88 | 82 | |
BC028288.1 | - | 1 - 2 | 185 | 4503501 | 82 | |
BC080758.1 | - | 1 - 2 | 100 | 5363932 | 82 | |
J02809.1 | - | 1 - 2 | 179 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC028288.1, BC080758.1, J02809.1, NM_008083.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
