Assay Details
| Gene Symbol: Ceacam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039185.1 | NP_001034274.1 | 4 - 5 | 1055 | 63 | |
| NM_001039186.1 | NP_001034275.1 | 4 - 5 | 1055 | 63 | ||
| GenBank mRNA | AB236332.2 | - | 4 - 5 | 964 | 63 | |
| AK085749.1 | - | 4 - 5 | 1015 | 63 | ||
| BC016891.1 | - | 4 - 5 | 1007 | 63 | ||
| BC121189.1 | - | 3 - 4 | 612 | 63 | ||
| M77196.1 | - | 4 - 5 | 1033 | 63 | ||
| X15351.1 | - | 4 - 5 | 1067 | 63 | ||
| X67279.1 | - | 4 - 5 | 1067 | 63 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Ceacam1
Genomic Map