Assay Details
| Gene Symbol: Nagpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013796.3 | NP_038824.2 | 3 - 4 | 716 | 64 | |
| NR_027804.1 | 2 - 3 | 813 | 64 | |||
| GenBank mRNA | AK032158.1 | - | 3 - 4 | 717 | 64 | |
| AK138035.1 | - | 3 - 4 | 709 | 64 | ||
| AK155450.1 | - | 3 - 4 | 695 | 64 | ||
| AK159629.1 | - | 3 - 4 | 695 | 64 | ||
| AK162633.1 | - | 3 - 4 | 698 | 64 | ||
| AK170219.1 | - | 3 - 4 | 698 | 64 | ||
| BC019640.1 | - | 2 - 3 | 778 | 64 | ||
| BC039790.1 | - | 3 - 4 | 708 | 4951805 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nagpa
Genomic Map