Assay Details
| Gene Symbol: Nagpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013796.3 | NP_038824.2 | 4 - 5 | 825 | 73 | |
| NR_027804.1 | 3 - 4 | 922 | 73 | |||
| GenBank mRNA | AK032158.1 | - | 4 - 5 | 826 | 73 | |
| AK138035.1 | - | 4 - 5 | 818 | 73 | ||
| AK155450.1 | - | 4 - 5 | 804 | 73 | ||
| AK159629.1 | - | 4 - 5 | 804 | 73 | ||
| AK162633.1 | - | 4 - 5 | 807 | 73 | ||
| AK168830.1 | - | 3 - 4 | 910 | 73 | ||
| AK170219.1 | - | 4 - 5 | 807 | 73 | ||
| BC019640.1 | - | 3 - 4 | 887 | 73 | ||
| BC039790.1 | - | 4 - 5 | 817 | 4951805 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nagpa
Genomic Map