Assay Details
| Gene Symbol: Nagpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013796.3 | NP_038824.2 | 8 - 9 | 1314 | 60 | |
| NR_027804.1 | 7 - 8 | 1411 | 60 | |||
| GenBank mRNA | AK032158.1 | - | 8 - 9 | 1315 | 60 | |
| AK087468.1 | - | 8 - 9 | 1317 | 60 | ||
| AK138035.1 | - | 8 - 9 | 1307 | 60 | ||
| AK155450.1 | - | 8 - 9 | 1293 | 60 | ||
| AK159629.1 | - | 8 - 9 | 1293 | 60 | ||
| AK162633.1 | - | 8 - 9 | 1297 | 60 | ||
| AK168830.1 | - | 7 - 8 | 1399 | 60 | ||
| AK170219.1 | - | 8 - 9 | 1296 | 60 | ||
| BC019640.1 | - | 7 - 8 | 1376 | 60 | ||
| BC039790.1 | - | 8 - 9 | 1306 | 4951805 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nagpa
Genomic Map