Assay Details
| Gene Symbol: Loxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010729.3 | NP_034859.2 | 2 - 3 | 1635 | 77 | |
| GenBank mRNA | AF357006.1 | - | 2 - 3 | 1396 | 77 | |
| BC003973.1 | - | 2 - 3 | 690 | 77 | ||
| BC037999.1 | - | 2 - 3 | 1619 | 5251968 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Loxl1
Genomic Map