Assay Details
| Gene Symbol: Loxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010729.3 | NP_034859.2 | 5 - 6 | 2030 | 70 | |
| GenBank mRNA | AF357006.1 | - | 5 - 6 | 1791 | 70 | |
| AK078512.1 | - | 4 - 5 | 1847 | 70 | ||
| BC003973.1 | - | 5 - 6 | 1085 | 70 | ||
| BC037999.1 | - | 5 - 6 | 2014 | 5251968 | 70 | |
| BC098243.1 | - | 2 - 3 | 613 | 70 | ||
| U79144.1 | - | 2 - 3 | 214 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Loxl1
Genomic Map