Assay Details
| Gene Symbol: Cd47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010581.3 | NP_034711.1 | 5 - 6 | 790 | 66 | |
| GenBank mRNA | AB012693.1 | - | 5 - 6 | 729 | 66 | |
| AK151922.1 | - | 4 - 5 | 673 | 66 | ||
| BC009094.1 | - | 3 - 4 | 356 | 66 | ||
| BC012667.1 | - | 5 - 6 | 724 | 4187965 | 66 | |
| HQ585874.1 | - | 4 - 5 | 594 | 66 | ||
| Z25524.1 | - | 5 - 6 | 790 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd47
Genomic Map