Assay Details
| Gene Symbol: Nde1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114085.1 | NP_001107557.1 | 1 - 2 | 97 | 65 | |
| NM_001285503.1 | NP_001272432.1 | 1 - 2 | 97 | 65 | ||
| NM_001285504.1 | NP_001272433.1 | 1 - 2 | 97 | 65 | ||
| NM_023317.2 | NP_075806.2 | 1 - 2 | 97 | 65 | ||
| GenBank mRNA | AK012861.1 | - | 1 - 2 | 47 | 65 | |
| AK150826.1 | - | 1 - 2 | 87 | 65 | ||
| AK166858.1 | - | 1 - 2 | 76 | 65 | ||
| BC023267.1 | - | 1 - 2 | 76 | 3672565 | 65 | |
| BY308148.1 | - | 1 - 2 | 98 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK150826.1, BC023267.1, NM_001114085.1, NM_001285503.1, NM_001285504.1, NM_023317.2 transcripts.
Set Membership: |
|

Nde1
Genomic Map