Assay Details
| Gene Symbol: Cct8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009840.3 | NP_033970.3 | 4 - 5 | 461 | 114 | |
| GenBank mRNA | AK010882.1 | - | 4 - 5 | 462 | 114 | |
| AK075711.1 | - | 4 - 5 | 442 | 114 | ||
| AK075880.1 | - | 4 - 5 | 450 | 114 | ||
| AK145661.1 | - | 4 - 5 | 445 | 114 | ||
| AK145754.1 | - | 4 - 5 | 444 | 114 | ||
| AK145918.1 | - | 4 - 5 | 463 | 114 | ||
| AK147038.1 | - | 4 - 5 | 464 | 114 | ||
| AK159969.1 | - | 4 - 5 | 453 | 114 | ||
| AK168465.1 | - | 4 - 5 | 433 | 114 | ||
| AK172867.1 | - | 4 - 5 | 432 | 114 | ||
| AK189477.1 | - | 3 - 4 | 314 | 114 | ||
| BC009007.1 | - | 4 - 5 | 419 | 3582639 | 114 | |
| Z37164.1 | - | 4 - 5 | 465 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cct8
Genomic Map