Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900007456.mm.1 |
NM_001033536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007456.mm.1 |
NM_001033536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007456.mm.1 |
TC0900000970.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007459.mm.1 |
NM_001033536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007459.mm.1 |
NM_001033536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007459.mm.1 |
TC0900000970.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007458.mm.1 |
NM_001033536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007458.mm.1 |
NM_001033536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007458.mm.1 |
TC0900000970.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007454.mm.1 |
NM_001033536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007454.mm.1 |
NM_001033536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007454.mm.1 |
TC0900000970.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007453.mm.1 |
NM_001033536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007453.mm.1 |
NM_001033536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007453.mm.1 |
TC0900000970.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007455.mm.1 |
NM_001033536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007455.mm.1 |
NM_001033536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007455.mm.1 |
TC0900000970.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007457.mm.1 |
NM_001033536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007457.mm.1 |
NM_001033536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007457.mm.1 |
TC0900000970.mm.1 |
MTA TC |