Assay Details
| Gene Symbol: Rfx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033536.1 | NP_001028708.1 | 4 - 5 | 983 | 72 | |
| GenBank mRNA | AK090370.1 | - | 4 - 5 | 984 | 72 | |
| AK170249.1 | - | 5 - 6 | 455 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfx7
Genomic Map