Assay Details
Gene Symbol: Dis3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001295.2 | NP_001001295.2 | 2 - 3 | 416 | 74 | |
NM_001177784.1 | NP_001171255.1 | 2 - 3 | 377 | 74 | ||
NM_172519.3 | NP_766107.1 | 2 - 3 | 320 | 74 | ||
GenBank mRNA | AK029974.1 | - | 2 - 3 | 321 | 74 | |
AK032451.1 | - | 2 - 3 | 325 | 74 | ||
AK035357.1 | - | 2 - 3 | 316 | 74 | ||
AK150851.1 | - | 2 - 3 | 366 | 74 | ||
AK151780.1 | - | 2 - 3 | 366 | 74 | ||
AK153433.1 | - | 2 - 3 | 366 | 74 | ||
AK173310.1 | - | 2 - 3 | 350 | 74 | ||
BC056939.1 | - | 2 - 3 | 349 | 5708685 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK029974.1, BC056939.1, NM_001177784.1, NM_172519.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
