Assay Details
| Gene Symbol: Zfp426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110309.1 | NP_001103779.1 | 1 - 2 | 47 | 60 | |
| GenBank mRNA | AK036912.1 | - | 1 - 2 | 35 | 60 | |
| BC023931.1 | - | 1 - 2 | 52 | 5324400 | 60 | |
| BC024805.1 | - | 1 - 2 | 43 | 5359610 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK036912.1, BC023931.1, BC024805.1, NM_001110309.1 transcripts.
Set Membership: |
|

Zfp426
Genomic Map