Assay Details
| Gene Symbol: Pde4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183408.3 | NP_899668.1 | 5 - 6 | 871 | 76 | |
| GenBank mRNA | AF038895.1 | - | 5 - 6 | 372 | 76 | |
| AK153657.1 | - | 5 - 6 | 875 | 76 | ||
| AK160843.1 | - | 4 - 5 | 305 | 76 | ||
| BC027224.1 | - | 5 - 6 | 567 | 76 | ||
| BC044786.2 | - | 4 - 5 | 1433 | 76 | ||
| BC139405.1 | - | 5 - 6 | 711 | 8862427 | 76 | |
| BC139408.1 | - | 5 - 6 | 711 | 8862430 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pde4a
Genomic Map