Assay Details
| Gene Symbol: Ccnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007630.2 | NP_031656.2 | 1 - 2 | 137 | 114 | |
| GenBank mRNA | AK013312.1 | - | 1 - 2 | 106 | 114 | |
| AK076122.1 | - | 1 - 2 | 131 | 114 | ||
| BY300844.1 | - | 1 - 2 | 140 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK013312.1, AK076122.1, NM_007630.2 transcripts.
Set Membership: |
|

Ccnb2
Genomic Map