Assay Details
| Gene Symbol: Dohh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133964.2 | NP_598725.2 | 2 - 3 | 475 | 69 | |
| GenBank mRNA | AK004052.1 | - | 2 - 3 | 441 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Dohh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133964.2 | NP_598725.2 | 2 - 3 | 475 | 69 | |
| GenBank mRNA | AK004052.1 | - | 2 - 3 | 441 | 69 | |
Set Membership: |
|