Assay Details
| Gene Symbol: Matk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285853.1 | NP_001272782.1 | 3 - 4 | 450 | 82 | |
| NM_001285854.1 | NP_001272783.1 | 3 - 4 | 447 | 82 | ||
| NM_001285855.1 | NP_001272784.1 | 3 - 4 | 515 | 82 | ||
| NM_010768.2 | NP_034898.1 | 4 - 5 | 583 | 82 | ||
| GenBank mRNA | AK079816.1 | - | 3 - 4 | 243 | 82 | |
| AK158883.1 | - | 3 - 4 | 308 | 82 | ||
| BC060086.1 | - | 3 - 4 | 404 | 82 | ||
| BC111864.1 | - | 3 - 4 | 261 | 40046807 | 82 | |
| D45243.1 | - | 3 - 4 | 515 | 82 | ||
| L27738.1 | - | 4 - 5 | 337 | 82 | ||
| S77473.1 | - | 3 - 4 | 262 | 82 | ||
| U05210.1 | - | 3 - 4 | 236 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Matk
Genomic Map