Assay Details
Gene Symbol: Lsm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025349.2 | NP_079625.1 | 2 - 3 | 121 | 63 | |
GenBank mRNA | AK003062.1 | - | 2 - 3 | 109 | 63 | |
AK007462.1 | - | 2 - 3 | 118 | 63 | ||
AK009317.1 | - | 2 - 3 | 108 | 63 | ||
AK012446.1 | - | 2 - 3 | 118 | 63 | ||
AK013355.1 | - | 2 - 3 | 122 | 63 | ||
BC027511.1 | - | 2 - 3 | 111 | 1382565 | 63 | |
BC081444.1 | - | 1 - 2 | 104 | 5711629 | 63 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
