Assay Details
Gene Symbol: Lrrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146048.1 | NP_001139520.1 | 2 - 3 | 498 | 64 | |
NM_172528.3 | NP_766116.3 | 2 - 3 | 146 | 64 | ||
GenBank mRNA | AK051043.1 | - | 2 - 3 | 497 | 64 | |
AK082721.1 | - | 2 - 3 | 147 | 64 | ||
AK147075.1 | - | 2 - 3 | 498 | 64 | ||
AK220166.1 | - | 2 - 3 | 516 | 64 | ||
BC046591.1 | - | 2 - 3 | 485 | 64 | ||
BC087542.1 | - | 2 - 3 | 345 | 4934980 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK082721.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
