Assay Details
Gene Symbol: Lrrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146048.1 | NP_001139520.1 | 5 - 6 | 726 | 125 | |
NM_172528.3 | NP_766116.3 | 5 - 6 | 374 | 125 | ||
GenBank mRNA | AK051043.1 | - | 5 - 6 | 725 | 125 | |
AK082721.1 | - | 5 - 6 | 375 | 125 | ||
AK147075.1 | - | 5 - 6 | 726 | 125 | ||
AK220166.1 | - | 5 - 6 | 744 | 125 | ||
BC046591.1 | - | 5 - 6 | 713 | 125 | ||
BC087542.1 | - | 5 - 6 | 573 | 4934980 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
