Assay Details
Gene Symbol: Fbxo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081490.2 | NP_001074959.1 | 11 - 12 | 1112 | 99 | |
NM_023605.2 | NP_076094.2 | 11 - 12 | 1414 | 99 | ||
GenBank mRNA | AB041585.1 | - | 11 - 12 | 1384 | 99 | |
AK077607.1 | - | 11 - 12 | 1075 | 99 | ||
BC020074.1 | - | 11 - 12 | 1385 | 3710611 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
